(2,6-Dichloro-3-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51321-500MG 51321-1G 51321-5G 51321-25G
77328-350EA 99.91 USD
77328-350 77342-812 77364-688 77376-186
(2,6-Dichloro-3-methoxyphenyl)(methyl)sulfane ≥97%
(2,6-Dichloro-3-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈Cl₂OS
MW: 223.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28360063
CAS Number: 1936630-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR