3,4-Difluoro-N,N,2-trimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

51287-1G 51287-5G 51287-25G
77341-922EA 108.15 USD
77341-922 77364-678 77376-170
3,4-Difluoro-N,N,2-trimethylbenzamide ≥97%
3,4-Difluoro-N,N,2-trimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₂NO
MW: 199.197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530085
CAS Number: 2734772-73-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR