(3,4-Difluoro-2-methylphenyl)(piperidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

51285-500MG 51285-1G 51285-5G 51285-25G
77328-342EA 85.49 USD
77328-342 77342-216 77364-674 77376-166
(3,4-Difluoro-2-methylphenyl)(piperidin-1-yl)methanone ≥97%
(3,4-Difluoro-2-methylphenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₅F₂NO
Storage Temperature: Ambient
MDL Number: MFCD34530083
CAS Number: 2734774-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR