(3,4-Difluoro-2-methylphenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

51284-500MG 51284-1G 51284-5G 51284-25G
77328-340EA 82.4 USD
77328-340 77342-150 77364-672 77376-164
(3,4-Difluoro-2-methylphenyl)(thiomorpholino)methanone ≥97%
(3,4-Difluoro-2-methylphenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₂NOS
MW: 257.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530082
CAS Number: 2734775-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR