(3,4-Difluoro-2-methylphenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

51283-500MG 51283-1G 51283-5G 51283-25G
77328-338EA 82.4 USD
77328-338 77342-148 77364-670 77376-162
(3,4-Difluoro-2-methylphenyl)(morpholino)methanone ≥97%
(3,4-Difluoro-2-methylphenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₂NO₂
MW: 241.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530081
CAS Number: 2734775-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR