3-Methoxy-2-methyl-6-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

51277-500MG 51277-1G 51277-25G 51277-5G
77328-334EA 301.79 USD
77328-334 77351-898 77386-890 77376-158
3-Methoxy-2-methyl-6-(methylthio)pyridine ≥95%
3-Methoxy-2-methyl-6-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₈H₁₁NOS
MW: 169.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564169
CAS Number: 2734774-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR