Methyl 4-chloro-3-ethoxy-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

51273-500MG 51273-1G 51273-5G 51273-25G
77328-332EA 100.94 USD
77328-332 77342-926 77364-666 77376-156
Methyl 4-chloro-3-ethoxy-2-fluorobenzoate ≥97%
Methyl 4-chloro-3-ethoxy-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD30717177
CAS Number: 2384635-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR