2'-(Hydroxymethyl)-[1,1'-biphenyl]-4-ol ≥97%

Supplier: AOBChem USA

Warning

51269-500MG 51269-1G 51269-5G
77336-038EA 374.92 USD
77336-038 77351-896 77376-154
2'-(Hydroxymethyl)-[1,1'-biphenyl]-4-ol ≥97%
2'-(Hydroxymethyl)-[1,1'-biphenyl]-4-ol

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18312795
CAS Number: 134967-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR