Methyl 3-isopropyl-5-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

51264-500MG 51264-1G 51264-5G
77338-136EA 466.59 USD
77338-136 77351-892 77376-150
Methyl 3-isopropyl-5-methoxybenzoate ≥97%
Methyl 3-isopropyl-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756773
CAS Number: 2119414-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR