3-Isopropyl-5-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

51261-500MG 51261-1G 51261-5G
77338-984EA 540.75 USD
77338-984 77351-890 77376-148
3-Isopropyl-5-methoxybenzaldehyde ≥95%
3-Isopropyl-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD28642207
CAS Number: 1204344-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR