2,6-Dichloro-3-isopropoxyphenol ≥97%

Supplier: AOBChem USA

Warning

51256-500MG 51256-1G 51256-5G 51256-25G
77328-328EA 208.06 USD
77328-328 77351-886 77364-664 77386-888
2,6-Dichloro-3-isopropoxyphenol ≥97%
2,6-Dichloro-3-isopropoxyphenol

Precaución: for Research Use Only

Formula: C₉H₁₀Cl₂O₂
MW: 221.081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564168
CAS Number: 2734779-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR