2,6-Dichloro-3-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

51251-500MG 51251-1G 51251-5G 51251-25G
77328-320EA 202.91 USD
77328-320 77351-880 77364-656 77386-882
2,6-Dichloro-3-isopropoxybenzoic acid ≥97%
2,6-Dichloro-3-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD33403963
CAS Number: 160647-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR