1,3-Dichloro-4-isopropoxy-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

51249-500MG 51249-1G 51249-5G
77337-882EA 450.11 USD
77337-882 77351-878 77376-142
1,3-Dichloro-4-isopropoxy-2-methylbenzene ≥95%
1,3-Dichloro-4-isopropoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂Cl₂O
MW: 219.108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403962
CAS Number: 2624417-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR