(2,6-Dichloro-3-isopropoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

51247-500MG 51247-1G 51247-5G 51247-25G
77328-318EA 161.71 USD
77328-318 77345-876 77364-654 77386-880
(2,6-Dichloro-3-isopropoxyphenyl)(methyl)sulfane ≥95%
(2,6-Dichloro-3-isopropoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₂Cl₂OS
MW: 251.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403960
CAS Number: 2624417-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR