1-(2,6-Dichloro-3-isopropoxyphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

51245-500MG 51245-1G 51245-5G 51245-25G
77328-316EA 181.28 USD
77328-316 77346-856 77364-652 77386-878
1-(2,6-Dichloro-3-isopropoxyphenyl)ethanol ≥97%
1-(2,6-Dichloro-3-isopropoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₁H₁₄Cl₂O₂
MW: 249.134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403959
CAS Number: 2624417-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR