Methyl 3-(benzyloxy)-2-fluoro-6-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

51232-500MG 51232-1G 51232-5G
77328-308EA 338.87 USD
77328-308 77351-870 77376-140
Methyl 3-(benzyloxy)-2-fluoro-6-methoxybenzoate ≥97%
Methyl 3-(benzyloxy)-2-fluoro-6-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₅FO₄
MW: 290.286 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530071
CAS Number: 2114573-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR