(3-(Benzyloxy)-2-fluoro-6-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

51230-500MG 51230-1G 51230-5G
77335-644EA 361.53 USD
77335-644 77351-866 77376-138
(3-(Benzyloxy)-2-fluoro-6-methoxyphenyl)methanol ≥95%
(3-(Benzyloxy)-2-fluoro-6-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₅H₁₅FO₃
MW: 262.276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564165
CAS Number: 1891645-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR