3-(Benzyloxy)-2-fluoro-6-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

51229-500MG 51229-1G 51229-25G 51229-5G
77328-304EA 304.88 USD
77328-304 77351-864 77386-868 77376-136
3-(Benzyloxy)-2-fluoro-6-methoxybenzaldehyde ≥97%
3-(Benzyloxy)-2-fluoro-6-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD34564164
CAS Number: 1895032-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR