1-(Benzyloxy)-3-chloro-2-fluoro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

51226-500MG 51226-1G 51226-5G 51226-25G
77328-300EA 160.68 USD
77328-300 77345-794 77364-640 77386-864
1-(Benzyloxy)-3-chloro-2-fluoro-4-methoxybenzene ≥97%
1-(Benzyloxy)-3-chloro-2-fluoro-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClFO₂
MW: 266.695 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902925
CAS Number: 2586126-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR