(3-(Benzyloxy)-2-fluoro-6-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51225-500MG 51225-1G 51225-5G 51225-25G
77328-298EA 177.16 USD
77328-298 77346-556 77364-638 77386-862
(3-(Benzyloxy)-2-fluoro-6-methoxyphenyl)(methyl)sulfane ≥97%
(3-(Benzyloxy)-2-fluoro-6-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₅H₁₅FO₂S
MW: 278.342 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902924
CAS Number: 2586126-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR