1-(3-(Benzyloxy)-2-fluoro-6-methoxyphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

51223-500MG 51223-1G 51223-5G
77339-118EA 552.08 USD
77339-118 77351-862 77376-134
1-(3-(Benzyloxy)-2-fluoro-6-methoxyphenyl)ethanol ≥95%
1-(3-(Benzyloxy)-2-fluoro-6-methoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₆H₁₇FO₃
MW: 276.303 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902923
CAS Number: 1897185-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR