5-(Benzyloxy)-1,3-difluoro-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

51219-500MG 51219-1G 51219-5G 51219-25G
77328-294EA 161.71 USD
77328-294 77345-874 77364-634 77386-860
5-(Benzyloxy)-1,3-difluoro-2-iodobenzene ≥97%
5-(Benzyloxy)-1,3-difluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉F₂IO
MW: 346.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902921
CAS Number: 1799824-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR