Ethyl 4-chloro-2-ethoxy-3-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

51198-500MG 51198-1G 51198-5G
77336-866EA 396.55 USD
77336-866 77351-854 77376-128
Ethyl 4-chloro-2-ethoxy-3-fluorobenzoate ≥97%
Ethyl 4-chloro-2-ethoxy-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClFO₃
MW: 246.663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530065
CAS Number: 2386387-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR