3-(Difluoromethoxy)-4-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

51190-500MG 51190-1G 51190-5G 51190-25G
77328-288EA 281.19 USD
77328-288 77351-850 77364-628 77386-854
3-(Difluoromethoxy)-4-fluorobenzoic acid ≥97%
3-(Difluoromethoxy)-4-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD14698510
CAS Number: 1214383-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR