1-(2,5-Difluoropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

51157-500MG 51157-1G
77340-408EA 1033.09 USD
77340-408 77351-844
1-(2,5-Difluoropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%
1-(2,5-Difluoropyridin-4-yl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₇H₂F₅NO
MW: 211.089 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28130479
CAS Number: 1866224-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR