1-(3-Chloro-2-fluoropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
51152-1G
51152-5G
51152-25G
77340-898EA
62.83
USD
77340-898
77364-622
77376-118
1-(3-Chloro-2-fluoropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%
1-(3-Chloro-2-fluoropyridin-4-yl)-2,2,2-trifluoroethanone
Precaución: for Research Use Only
Formula:
C₇H₂ClF₄NO Storage Temperature: Ambient |
MDL Number:
MFCD28130421 CAS Number: 1824270-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...