1-(2,3-Dichloropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

51151-1G 51151-5G 51151-25G
77341-106EA 71.07 USD
77341-106 77364-620 77376-116
1-(2,3-Dichloropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%
1-(2,3-Dichloropyridin-4-yl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₇H₂Cl₂F₃NO
MW: 243.998 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22381702
CAS Number: 1375303-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR