4-Fluoro-2,6-dinitroaniline ≥95%

Supplier: AOBChem USA

Warning

51149-1G 51149-5G 51149-25G
77341-738EA 103 USD
77341-738 77364-618 77376-114
4-Fluoro-2,6-dinitroaniline ≥95%
4-Fluoro-2,6-dinitroaniline

Precaución: for Research Use Only

Formula: C₆H₄FN₃O₄
Storage Temperature: Ambient
MDL Number: MFCD24160434
CAS Number: 82366-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR