1-(3,5-Dichloropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

51139-500MG 51139-1G 51139-5G
77339-754EA 628.3 USD
77339-754 77351-842 77376-112
1-(3,5-Dichloropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%
1-(3,5-Dichloropyridin-4-yl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₇H₂Cl₂F₃NO
Storage Temperature: Ambient
CAS Number: 2228452-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR