1-(3-Chloropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

51126-500MG 51126-1G 51126-5G 51126-25G
77328-282EA 247.2 USD
77328-282 77351-840 77364-616 77386-848
1-(3-Chloropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%
1-(3-Chloropyridin-4-yl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO
MW: 209.553 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189040
CAS Number: 1060802-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR