2,2,2-Trifluoro-1-(5-methoxypyridin-3-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

50895-500MG 50895-1G 50895-5G
77339-752EA 628.3 USD
77339-752 77351-838 77376-108
2,2,2-Trifluoro-1-(5-methoxypyridin-3-yl)ethanone ≥97%
2,2,2-Trifluoro-1-(5-methoxypyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₂
MW: 205.134 g/mol
Storage Temperature: Ambient
CAS Number: 1060801-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR