Methyl 5-bromo-2-fluoro-3-iodobenzoate ≥97%

Supplier: AOBChem USA

Warning

50806-500MG 50806-1G 50806-5G
77335-540EA 357.41 USD
77335-540 77351-832 77376-104
Methyl 5-bromo-2-fluoro-3-iodobenzoate ≥97%
Methyl 5-bromo-2-fluoro-3-iodobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrFIO₂
MW: 358.931 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24107100
CAS Number: 1510575-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR