(5-Bromo-2-fluoro-3-iodophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

50804-500MG 50804-1G 50804-5G
77339-030EA 545.9 USD
77339-030 77351-828 77376-102
(5-Bromo-2-fluoro-3-iodophenyl)methanol ≥97%
(5-Bromo-2-fluoro-3-iodophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅BrFIO
MW: 330.921 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530061
CAS Number: 2090447-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR