(5-Bromo-2-fluoro-3-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

50801-500MG 50801-1G 50801-5G 50801-25G
77328-274EA 184.37 USD
77328-274 77347-208 77364-606 77386-840
(5-Bromo-2-fluoro-3-iodophenyl)(methyl)sulfane ≥97%
(5-Bromo-2-fluoro-3-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅BrFIS
Storage Temperature: Ambient
MDL Number: MFCD33403957
CAS Number: 2385051-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR