1-(5-Bromo-2-fluoro-3-iodophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

50800-500MG 50800-1G 50800-5G 50800-25G
77328-272EA 116.39 USD
77328-272 77343-418 77364-604 77376-100
1-(5-Bromo-2-fluoro-3-iodophenyl)ethanone ≥97%
1-(5-Bromo-2-fluoro-3-iodophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₅BrFIO
Storage Temperature: Ambient
MDL Number: MFCD34756757
CAS Number: 2092862-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR