2-(5-Bromo-2-fluoro-3-iodophenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

50798-500MG 50798-1G 50798-5G 50798-25G
77328-270EA 126.69 USD
77328-270 77344-120 77364-602 77376-098
2-(5-Bromo-2-fluoro-3-iodophenyl)propan-2-ol ≥97%
2-(5-Bromo-2-fluoro-3-iodophenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₉H₉BrFIO
MW: 358.974 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178598
CAS Number: 2643368-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR