5-Bromo-2-fluoro-[1,1'-biphenyl]-3-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

50788-500MG 50788-1G 50788-5G
77338-702EA 510.88 USD
77338-702 77351-822 77376-092
5-Bromo-2-fluoro-[1,1'-biphenyl]-3-carbaldehyde ≥95%
5-Bromo-2-fluoro-[1,1'-biphenyl]-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₈BrFO
MW: 279.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902908
CAS Number: 2586127-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR