3-Chloro-2-(methylthio)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%

Supplier: AOBChem USA

Warning

50786-500MG 50786-1G 50786-5G 50786-25G
77328-268EA 181.28 USD
77328-268 77346-854 77364-600 77386-838
3-Chloro-2-(methylthio)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%
3-Chloro-2-(methylthio)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₇BClNO₂S
MW: 285.598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29116631
CAS Number: 1351229-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR