5-Chloro-6-(methylthio)nicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

50783-500MG 50783-1G 50783-25G 50783-5G
77328-266EA 284.28 USD
77328-266 77351-820 77386-836 77376-090
5-Chloro-6-(methylthio)nicotinaldehyde ≥97%
5-Chloro-6-(methylthio)nicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆ClNOS
MW: 187.647 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29125960
CAS Number: 1288998-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR