Ethyl 2-fluoro-3,4-dimethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

50778-500MG 50778-1G 50778-5G 50778-25G
77328-262EA 217.33 USD
77328-262 77351-818 77364-596 77386-834
Ethyl 2-fluoro-3,4-dimethoxybenzoate ≥97%
Ethyl 2-fluoro-3,4-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₄
MW: 228.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19439846
CAS Number: 1806390-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR