2-Fluoro-1-iodo-3,4-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

50773-500MG 50773-1G 50773-5G 50773-25G
77328-258EA 141.11 USD
77328-258 77344-816 77364-592 77386-830
2-Fluoro-1-iodo-3,4-dimethoxybenzene ≥97%
2-Fluoro-1-iodo-3,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈FIO₂
MW: 282.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28793038
CAS Number: 1803783-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR