(4-Fluoro-3-isopropoxy-5-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

50770-500MG 50770-1G 50770-5G
77336-074EA 375.95 USD
77336-074 77351-814 77376-086
(4-Fluoro-3-isopropoxy-5-methylphenyl)methanol ≥97%
(4-Fluoro-3-isopropoxy-5-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₅FO₂
MW: 198.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530060
CAS Number: 2734776-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR