Methyl 4-fluoro-3-isopropoxy-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

50769-500MG 50769-1G 50769-5G
77328-256EA 345.05 USD
77328-256 77351-812 77376-084
Methyl 4-fluoro-3-isopropoxy-5-methylbenzoate ≥97%
Methyl 4-fluoro-3-isopropoxy-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅FO₃
Storage Temperature: Ambient
MDL Number: MFCD34178594
CAS Number: 2643368-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR