Ethyl 4-fluoro-3-isopropoxy-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

50768-500MG 50768-1G 50768-25G 50768-5G
77328-254EA 289.43 USD
77328-254 77351-810 77386-828 77376-082
Ethyl 4-fluoro-3-isopropoxy-5-methylbenzoate ≥97%
Ethyl 4-fluoro-3-isopropoxy-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₇FO₃
MW: 240.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178593
CAS Number: 2643368-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR