3-Fluoro-2-iodo-5-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

50738-500MG 50738-1G 50738-5G 50738-25G
77328-252EA 281.19 USD
77328-252 77351-800 77364-590 77386-826
3-Fluoro-2-iodo-5-(trifluoromethyl)phenol ≥97%
3-Fluoro-2-iodo-5-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₃F₄IO
MW: 305.996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756743
CAS Number: 2385779-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR