3-Fluoro-2-methyl-5-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

50736-500MG 50736-1G 50736-5G
77328-250EA 332.69 USD
77328-250 77351-798 77376-072
3-Fluoro-2-methyl-5-(trifluoromethyl)phenol ≥97%
3-Fluoro-2-methyl-5-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₈H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD34530054
CAS Number: 2385454-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR