(2-Fluoro-3-iodo-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

50732-500MG 50732-1G 50732-5G 50732-25G
77328-248EA 122.57 USD
77328-248 77343-926 77364-588 77376-070
(2-Fluoro-3-iodo-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(2-Fluoro-3-iodo-5-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₅F₄IS
MW: 336.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756741
CAS Number: 2705758-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR