1-Fluoro-2-iodo-3-methoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

50711-500MG 50711-1G 50711-5G 50711-25G
77328-246EA 95.79 USD
77328-246 77342-618 77364-586 77376-066
1-Fluoro-2-iodo-3-methoxy-4-methylbenzene ≥97%
1-Fluoro-2-iodo-3-methoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈FIO
MW: 266.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902905
CAS Number: 2383375-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR