2-Bromo-1-fluoro-3-methoxy-4-methylbenzene ≥95%
Supplier: AOBChem USA
|
Warning
|
50709-500MG
50709-1G
50709-25G
50709-5G
77328-244EA
304.88
USD
77328-244
77351-794
77386-824
77376-064
2-Bromo-1-fluoro-3-methoxy-4-methylbenzene ≥95%
2-Bromo-1-fluoro-3-methoxy-4-methylbenzene
Precaución: for Research Use Only
Formula:
C₈H₈BrFO Storage Temperature: Ambient |
MDL Number:
MFCD22414747 CAS Number: 1780793-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...