1-Fluoro-3-methoxy-2,4-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

50708-500MG 50708-1G 50708-5G 50708-25G
77328-242EA 120.51 USD
77328-242 77343-736 77364-584 77376-062
1-Fluoro-3-methoxy-2,4-dimethylbenzene ≥95%
1-Fluoro-3-methoxy-2,4-dimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD28794223
CAS Number: 26829-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR